Skip to Content
MilliporeSigma
Naratriptan hydrochloride ≥98% (HPLC)

Naratriptan hydrochloride

Naratriptan hydrochloride ≥98% (HPLC)
Synonym(s):
Amerge, GR-85548A, N-Methyl-3-(1-methyl-4-piperidinyl)-1H-indole-5-ethanesulfonamide hydrochloride
Empirical Formula (Hill Notation):
C17H25N3O2S ·HCl
CAS Number:
Molecular Weight:
371.93

Products

Product Category
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2