Skip to Content
MilliporeSigma
Naphthol AS-BI β-D-glucuronide β-glucuronidase substrate

Naphthol AS-BI β-D-glucuronide

Naphthol AS-BI β-D-glucuronide β-glucuronidase substrate
Synonym(s):
O-β-D-Glucuronosyl-naphthol AS-BI
Empirical Formula (Hill Notation):
C24H22BrNO9
CAS Number:
Molecular Weight:
548.34

Products

Product Category
Brand
Formula Weight
Physical Form
Showing 1-1 of 1