Skip to Content
MilliporeSigma
N-Acetylputrescine hydrochloride ≥98% (TLC)

N-Acetylputrescine hydrochloride

N-Acetylputrescine hydrochloride ≥98% (TLC)
Empirical Formula (Hill Notation):
C6H14N2O · HCl
CAS Number:
Molecular Weight:
166.65

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing