Skip to Content
MilliporeSigma
N-Acetylglycine ReagentPlus®, 99%

N-Acetylglycine

N-Acetylglycine ReagentPlus®, 99%
Synonym(s):
Acetamidoacetic acid, Aceturic acid
Linear Formula:
CH3CONHCH2CO2H
CAS Number:
Molecular Weight:
117.10
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing