Skip to Content
MilliporeSigma
N-Acetyl-D-penicillamine ≥99.0% (T)

N-Acetyl-D-penicillamine

N-Acetyl-D-penicillamine ≥99.0% (T)
Synonym(s):
N-Acetyl-3-mercapto-D-valine
Empirical Formula (Hill Notation):
C7H13NO3S
CAS Number:
Molecular Weight:
191.25
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing