Skip to Content
MilliporeSigma
N-Acetyl-D-mannosamine ≥98% (TLC)

N-Acetyl-D-mannosamine

N-Acetyl-D-mannosamine ≥98% (TLC)
Empirical Formula (Hill Notation):
C8H15NO6
CAS Number:
Molecular Weight:
221.21

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing