Skip to Content
MilliporeSigma
N-Acetyl-Asp-Glu ≥97% (TLC)

N-Acetyl-Asp-Glu

N-Acetyl-Asp-Glu ≥97% (TLC)
Synonym(s):
NAAG, Spaglumic acid
Empirical Formula (Hill Notation):
C11H16N2O8
CAS Number:
Molecular Weight:
304.25

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing