Skip to Content
MilliporeSigma
Nα-Benzoyl-L-arginine 4-nitroanilide hydrochloride >98% (TLC)

Nα-Benzoyl-L-arginine 4-nitroanilide hydrochloride

Nα-Benzoyl-L-arginine 4-nitroanilide hydrochloride >98% (TLC)
Synonym(s):
L-BAPA, L-BAPNA, BANI
Empirical Formula (Hill Notation):
C19H22N6O4 · HCl
CAS Number:
Molecular Weight:
434.88
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment