Skip to Content
MilliporeSigma
N-(5-Fluoresceinyl)maleimide suitable for fluorescence, ≥90% (HPLC), BioReagent

N-(5-Fluoresceinyl)maleimide

N-(5-Fluoresceinyl)maleimide suitable for fluorescence, ≥90% (HPLC), BioReagent
Synonym(s):
5-Maleimido-fluorescein
Empirical Formula (Hill Notation):
C24H13NO7
CAS Number:
Molecular Weight:
427.36

Products

Product Category
Brand
Formula Weight
Product Line
Purity
Showing 1-1 of 1