Skip to Content
MilliporeSigma
N-(2-Aminoethyl)acetamide technical grade, 90%

N-(2-Aminoethyl)acetamide

N-(2-Aminoethyl)acetamide technical grade, 90%
Synonym(s):
N-Acetylethylenediamine
Linear Formula:
CH3C(O)NHCH2CH2NH2
CAS Number:
Molecular Weight:
102.14

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing