Skip to Content
MilliporeSigma
Myxothiazol from Myxococcus fulvus Mx f85, ≥98% (HPLC)

Myxothiazol

Myxothiazol from Myxococcus fulvus Mx f85, ≥98% (HPLC)
Empirical Formula (Hill Notation):
C25H33N3O3S2
CAS Number:
Molecular Weight:
487.68

Products

Product Category
Brand
Biological Source
Formula Weight
Purity
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1