![m-Toluidine 99%](/deepweb/assets/sigmaaldrich/product/structures/423/104/97e45590-8e3f-49ad-b57e-f3ca96dae348/640/97e45590-8e3f-49ad-b57e-f3ca96dae348.png)
m-Toluidine
![m-Toluidine 99%](/deepweb/assets/sigmaaldrich/product/structures/423/104/97e45590-8e3f-49ad-b57e-f3ca96dae348/640/97e45590-8e3f-49ad-b57e-f3ca96dae348.png)
Synonym(s):
3-Methylaniline, m-Toluidine
Linear Formula:
3-(CH3)C6H4NH2
CAS Number:
Molecular Weight:
107.15
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing