Skip to Content
MilliporeSigma
MOPS hemisodium salt ≥99% (titration)

MOPS hemisodium salt

MOPS hemisodium salt ≥99% (titration)
Synonym(s):
3-(N-Morpholino)propanesulfonic acid hemisodium salt, 4-Morpholinepropanesulfonic acid hemisodium salt
Empirical Formula (Hill Notation):
C7H14.5NO4S · 0.5Na
CAS Number:
Molecular Weight:
220.25

Products

Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
pK Value
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing