Skip to Content
MilliporeSigma
Monoclonal Anti-CD71 antibody produced in mouse clone MEM-75, purified immunoglobulin, buffered aqueous solution

Monoclonal Anti-CD71 antibody produced in mouse

Monoclonal Anti-CD71 antibody produced in mouse clone MEM-75, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-TFRC, Anti-Transferrin Receptor

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique