Skip to Content
MilliporeSigma
Monensin sodium salt 90-95% (TLC)

Monensin sodium salt

Monensin sodium salt 90-95% (TLC)
Synonym(s):
Monensin A sodium salt
Empirical Formula (Hill Notation):
C36H61NaO11
CAS Number:
Molecular Weight:
692.85
EC Number:

Products

Product Category
Brand
Formula Weight
Purity
Activity Spectrum
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing