Skip to Content
MilliporeSigma
Mirtazapine Related Compound D certified reference material, pharmaceutical secondary standard

Mirtazapine Related Compound D

Mirtazapine Related Compound D certified reference material, pharmaceutical secondary standard
Synonym(s):
1,3,4,14b-Tetrahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2]benzazepin-10(2H)-one, 10-Oxomirtazapine
Empirical Formula (Hill Notation):
C17H17N3O
CAS Number:
Molecular Weight:
279.34

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade