Skip to Content
MilliporeSigma
Metoclopramide Impurity C Pharmaceutical Secondary Standard; Certified Reference Material

Metoclopramide Impurity C

Metoclopramide Impurity C Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
4-Amino-5-chloro-2-methoxybenzoic acid, 4-Amino-5-chloro-o-anisic acid
Linear Formula:
H2NC6H2(Cl)(OCH3)CO2H
CAS Number:
Molecular Weight:
201.61

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1