Skip to Content
MilliporeSigma
Methyltriphenoxyphosphonium iodide 96%

Methyltriphenoxyphosphonium iodide

Methyltriphenoxyphosphonium iodide 96%
Synonym(s):
Triphenyl phosphite methoiodide
Linear Formula:
CH3P(I)(OC6H5)3
CAS Number:
Molecular Weight:
452.22
EC Number:

Products

Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing