Skip to Content
MilliporeSigma
(−)-Methyl (S)-2,2-dimethyl-1,3-dioxolane-4-carboxylate 96%

(−)-Methyl (S)-2,2-dimethyl-1,3-dioxolane-4-carboxylate

(−)-Methyl (S)-2,2-dimethyl-1,3-dioxolane-4-carboxylate 96%
Synonym(s):
α,β-Isopropylidene-L-glyceric acid methyl ester, Methyl α,β-isopropylidene-L-glycerate, Methyl 2,3-O-isopropylidene-L-glycerate
Empirical Formula (Hill Notation):
C7H12O4
CAS Number:
Molecular Weight:
160.17

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing