Skip to Content
MilliporeSigma
Methyl propyl trisulfide 50%, FG

Methyl propyl trisulfide

Methyl propyl trisulfide 50%, FG
Empirical Formula (Hill Notation):
C4H10S3
CAS Number:
Molecular Weight:
154.32

Products

Product Category
Brand
Biological Source
Formula Weight
Agency
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing