Skip to Content
MilliporeSigma
Methyl pentadecanoate analytical standard

Methyl pentadecanoate

Methyl pentadecanoate analytical standard
Synonym(s):
Pentadecanoic acid methyl ester
Linear Formula:
CH3(CH2)13CO2CH3
CAS Number:
Molecular Weight:
256.42
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing