Skip to Content
MilliporeSigma
vibrant-m

Methylnitrosoindoline

vibrant-m
Synonym(s):
2,3-Dihydro-2-methyl-1-nitroso-1H-indole
Empirical Formula (Hill Notation):
C9H10N2O
CAS Number:
Molecular Weight:
162.19
EC Number:

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing