Skip to Content
MilliporeSigma
Methylmalonic acid analytical standard

Methylmalonic acid

Methylmalonic acid analytical standard
Synonym(s):
2-Methylpropanedioic acid
Linear Formula:
CH3CH(CO2H)2
CAS Number:
Molecular Weight:
118.09

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing