Skip to Content
MilliporeSigma
Methylguanidine hydrochloride 98%

Methylguanidine hydrochloride

Methylguanidine hydrochloride 98%
Linear Formula:
CH3NHC(=NH)NH2·HCl
CAS Number:
Molecular Weight:
109.56

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing