Skip to Content
MilliporeSigma
Methylene Blue certified reference material, pharmaceutical secondary standard

Methylene blue

Methylene Blue certified reference material, pharmaceutical secondary standard
Synonym(s):
3,7-bis(Dimethylamino)phenazathionium chloride, Tetramethylthionine chloride, Methylene Blue hydrate
Empirical Formula (Hill Notation):
C16H18ClN3S · xH2O
CAS Number:
Molecular Weight:
319.85 (anhydrous basis)

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
Special Grade