Skip to Content
MilliporeSigma
Methyl dihydrojasmonate, mixture of cis and trans analytical standard

Methyl dihydrojasmonate, mixture of cis and trans

Methyl dihydrojasmonate, mixture of cis and trans analytical standard
Synonym(s):
(3-Oxo-2-pentylcyclopentyl)acetic acid methyl ester, Methyl (3-oxo-2-pentylcyclopentyl)acetate
Empirical Formula (Hill Notation):
C13H22O3
CAS Number:
Molecular Weight:
226.31
EC Number:

Products

Product Category
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade