Skip to Content
MilliporeSigma
Methylammonium bromide ≥99%, anhydrous

Methylammonium bromide

Methylammonium bromide ≥99%, anhydrous
Empirical Formula (Hill Notation):
CH6BrN
CAS Number:
Molecular Weight:
111.97

Products

Product Category
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing