Skip to Content
MilliporeSigma
Methyl acetimidate hydrochloride technical grade

Methyl acetimidate hydrochloride

Methyl acetimidate hydrochloride technical grade
Linear Formula:
CH3C(=NH)OCH3 · HCl
CAS Number:
Molecular Weight:
109.55
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing