Skip to Content
MilliporeSigma
Methyl 4-pentenoate ≥95.0% (GC)

Methyl 4-pentenoate

Methyl 4-pentenoate ≥95.0% (GC)
Synonym(s):
4-Pentenoic acid methyl ester, Allylacetic acid methyl ester, Methyl allylacetate
Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14

Products

Product Category
Brand
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing