Skip to Content
MilliporeSigma
Methyl 4-nitrobutyrate 97%

Methyl 4-nitrobutyrate

Methyl 4-nitrobutyrate 97%
Synonym(s):
4-Nitrobutyric acid methyl ester
Linear Formula:
O2N(CH2)3CO2CH3
CAS Number:
Molecular Weight:
147.13
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing