Skip to Content
MilliporeSigma
Methyl 4-methoxybutyrate 98%

Methyl 4-methoxybutyrate

Methyl 4-methoxybutyrate 98%
Linear Formula:
CH3O(CH2)3CO2CH3
CAS Number:
Molecular Weight:
132.16

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing