Skip to Content
MilliporeSigma
Methyl 4-hydroxyphenylacetate ReagentPlus®, 99%

Methyl 4-hydroxyphenylacetate

Methyl 4-hydroxyphenylacetate ReagentPlus®, 99%
Linear Formula:
HOC6H4CH2CO2CH3
CAS Number:
Molecular Weight:
166.17
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing