Skip to Content
MilliporeSigma
Methyl 4-formylbenzoate 99%

Methyl 4-formylbenzoate

Methyl 4-formylbenzoate 99%
Synonym(s):
Methyl benzaldehyde-4-carboxylate, Methyl terephthalaldehydate, Methyl 4-formylbenzoate
Linear Formula:
4-(OHC)C6H4COOCH3
CAS Number:
Molecular Weight:
164.16

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing