Skip to Content
MilliporeSigma
Methyl 4-(bromomethyl)benzoate 98%

Methyl 4-(bromomethyl)benzoate

Methyl 4-(bromomethyl)benzoate 98%
Linear Formula:
BrCH2C6H4CO2CH3
CAS Number:
Molecular Weight:
229.07

Products

Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing