Skip to Content
MilliporeSigma
Methyl 4-bromo-2-methoxybenzoate 98%

Methyl 4-bromo-2-methoxybenzoate

Methyl 4-bromo-2-methoxybenzoate 98%
Linear Formula:
BrC6H3(OCH3)CO2CH3
CAS Number:
Molecular Weight:
245.07

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing