Skip to Content
MilliporeSigma
Methyl 3-(methylthio)propionate 98%

Methyl 3-(methylthio)propionate

Methyl 3-(methylthio)propionate 98%
Synonym(s):
Methyl 3-(methylmercapto)propionate
Linear Formula:
CH3SCH2CH2COOCH3
CAS Number:
Molecular Weight:
134.20
EC Number:

Products

Product Category
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing