Skip to Content
MilliporeSigma
Methyl 2-oxobutanoate 95%

Methyl 2-oxobutanoate

Methyl 2-oxobutanoate 95%
Synonym(s):
2-Oxobutyric acid methyl ester, Methyl 2-ketobutyrate
Empirical Formula (Hill Notation):
C5H8O3
CAS Number:
Molecular Weight:
116.12

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing