Skip to Content
MilliporeSigma
Methyl 2-methyl-3-furyl disulfide ≥98%, FG

Methyl 2-methyl-3-furyl disulfide

Methyl 2-methyl-3-furyl disulfide ≥98%, FG
Empirical Formula (Hill Notation):
C6H8OS2
CAS Number:
Molecular Weight:
160.26
EC Number:

Products

Product Category
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing