Skip to Content
MilliporeSigma
Methyl 12-hydroxystearate European Pharmacopoeia (EP) Reference Standard

Methyl 12-hydroxystearate

Methyl 12-hydroxystearate European Pharmacopoeia (EP) Reference Standard
Synonym(s):
12-Hydroxystearic acid methyl ester, Methyl 12-hydroxyoctadecanoate
Empirical Formula (Hill Notation):
C19H38O3
CAS Number:
Molecular Weight:
314.50

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Format
Functional Group
Application
API Family
Reference Material Type
Special Grade