Skip to Content
MilliporeSigma
Methscopolamine Bromide pharmaceutical secondary standard, certified reference material

Methscopolamine bromide

Methscopolamine Bromide pharmaceutical secondary standard, certified reference material
Synonym(s):
(−)-Scopolamine methyl bromide, Hyoscine methyl bromide, Methscopolamine bromide
Empirical Formula (Hill Notation):
C18H24NO4 · Br
CAS Number:
Molecular Weight:
398.29

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade