Skip to Content
MilliporeSigma
Methoxyperfluorobutane 99%, mixture of n- and iso-butyl isomers

Methoxyperfluorobutane

Methoxyperfluorobutane 99%, mixture of n- and iso-butyl isomers
Synonym(s):
Nonafluorobutyl methyl ether, HFE-7100, Methoxynonafluorobutane, Methoxyperfluorobutane, Nonafluoromethoxybutane
Linear Formula:
CH3OCF2R, R = -CF2CF2CF3 or -CF(CF3)2
CAS Number:
Molecular Weight:
250.06

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing