Skip to Content
MilliporeSigma
Methanesulfonic acid concentrate 0.1 M CH3SO3H in water (0.1N), eluent concentrate for IC

Methanesulfonic acid concentrate

Methanesulfonic acid concentrate 0.1 M CH3SO3H in water (0.1N), eluent concentrate for IC
Synonym(s):
Methanesulfonic acid solution
Linear Formula:
CH3SO3H
CAS Number:
Molecular Weight:
96.11

Products

Brand
Formula Weight
Quality Segment
Markush Class
Markush Group
Technique
Showing 1-1 of 1