Skip to Content
MilliporeSigma
Metaxalone Related Compound B certified reference material, pharmaceutical secondary standard

Metaxalone Related Compound B

Metaxalone Related Compound B certified reference material, pharmaceutical secondary standard
Synonym(s):
1-Amino-3-(3,5-dimethylphenoxy)propan-2-ol
Empirical Formula (Hill Notation):
C11H17NO2
CAS Number:
Molecular Weight:
195.26

Products

Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade