Skip to Content
MilliporeSigma
Metaraminol (+)-bitartrate salt

Metaraminol (+)-bitartrate salt

Metaraminol (+)-bitartrate salt
Synonym(s):
(−)-m-Hydroxyphenylpropanolamine bitartrate salt
Empirical Formula (Hill Notation):
C9H13NO2 · C4H6O6
CAS Number:
Molecular Weight:
317.29
EC Number:

Products

Product Category
Formula Weight
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing