Skip to Content
MilliporeSigma
MES hemisodium salt free-flowing, Redi-Dri™, ≥98%

MES hemisodium salt

MES hemisodium salt free-flowing, Redi-Dri™, ≥98%
Synonym(s):
2-(N-Morpholino)ethanesulfonic acid hemisodium salt, 4-Morpholineethanesulfonic acid hemisodium salt
Empirical Formula (Hill Notation):
C6H13NO4S·0.5Na
CAS Number:
Molecular Weight:
206.73

Products

Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
pK Value
Product Line
Purity
Quality Segment
Showing 1-3 of 3