![Mercury(II) iodide red puriss. p.a., ACS reagent, red, ≥99.0% (RT)](/deepweb/assets/sigmaaldrich/product/structures/285/869/8fe1a00a-d88f-4e52-a834-dd77664bf2d4/640/8fe1a00a-d88f-4e52-a834-dd77664bf2d4.png)
Mercury(II) iodide red
![Mercury(II) iodide red puriss. p.a., ACS reagent, red, ≥99.0% (RT)](/deepweb/assets/sigmaaldrich/product/structures/285/869/8fe1a00a-d88f-4e52-a834-dd77664bf2d4/640/8fe1a00a-d88f-4e52-a834-dd77664bf2d4.png)
Synonym(s):
Mercuric iodide red, Mercury(II) iodide red,
Empirical Formula (Hill Notation):
HgI2
CAS Number:
Molecular Weight:
454.40
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing