Skip to Content
MilliporeSigma
Mercury(II) iodide ACS reagent, ≥99.0%

Mercury(II) iodide

Mercury(II) iodide ACS reagent, ≥99.0%
Synonym(s):
Mercuric iodide red, Mercury(II) iodide
Linear Formula:
HgI2
CAS Number:
Molecular Weight:
454.40

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Reagent Type
Core
Special Grade
Showing 1-2 of 2