Skip to Content
MilliporeSigma
m-Cresol Purple sodium salt Dye content 90 %

m-Cresol Purple sodium salt

m-Cresol Purple sodium salt Dye content 90 %
Synonym(s):
m-Cresolsulfonphthalein sodium salt
Empirical Formula (Hill Notation):
C21H17NaO5S
CAS Number:
Molecular Weight:
404.41
EC Number:

Products

Product Category
Color
Formula Weight
Physical Form
Chemical Composition
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing