Skip to Content
MilliporeSigma
Manganese(III) acetylacetonate technical grade

Manganese(III) acetylacetonate

Manganese(III) acetylacetonate technical grade
Synonym(s):
2,4-Pentanedione manganese(III) derivative, Manganic acetylacetonate, Mn(acac)3
Linear Formula:
Mn(C5H7O2)3
CAS Number:
Molecular Weight:
352.26
EC Number:

Products

Formula Weight
Melting Point (°C)
Markush Class
Markush Group
Reagent Type
Core
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing