Skip to Content
MilliporeSigma
Manganese(II) chloride bis(lithium chloride) complex solution 0.5 M in THF, density: 0.956 g/mL at 25 °C

Manganese(II) chloride bis(lithium chloride) complex solution

Manganese(II) chloride bis(lithium chloride) complex solution 0.5 M in THF, density: 0.956 g/mL at 25 °C
Synonym(s):
Lithium tetrachloromanganate(II) solution, Manganese(II) chloride bis(lithium chloride) complex solution
Linear Formula:
Li2MnCl4
CAS Number:
Molecular Weight:
210.63

Products

Brand
Formula Weight
Physical Form
Quality Segment
Reagent Type
Core